Learn More
Medchemexpress LLC HA/Hemagglutinin H1 20ug
SDP
Catalog No. 5000179887
Encompass_Preferred
HA/Hemagglutinin Protein is a viral surface glycoprotein that mediates virus adsorption to host cells and promotes membrane fusion required for infection HA/Hemagglutinin Protein plays a key role in the process of viral infection of host cells HA/Hemagglutinin Protein H1N1 (Biotinylated ACP41953 1 HEK293 His) is a recombinant HA/Hemagglutinin protein expressed by HEK293 and carrying a C-His tag[1][2]
Catalog No. 50-001-79887 Supplier Medchemexpress LLC Supplier No. HYP7403820UG
HA/Hemagglutinin Protein is a viral surface glycoprotein that mediates virus adsorption to host cells and promotes membrane fusion required for infection HA/Hemagglutinin Protein plays a key role in the process of viral infection of host cells HA/Hemagglutinin Protein H1N1 (Biotinylated ACP41953 1 HEK293 His) is a recombinant HA/Hemagglutinin protein expressed by HEK293 and carrying a C-His tag[1][2]