Learn More
Medchemexpress LLC HA/Hemagglutinin H1 5ug
SDP
Supplier:  Medchemexpress LLC HYP740345UG
Encompass_Preferred
Hemagglutinin protein (HA) is a class I viral fusion protein that binds to sialic acid-containing receptors and initiates viral attachment to cells This binding promotes internalization of virions through various endocytic pathways HA/Hemagglutinin Protein H1N1 (P03452 sf9 His) is the recombinant Virus-derived HA/Hemagglutinin protein expressed by Sf9 insect cells with C-His labeled tag
Catalog No. 50-001-79090