Learn More
Medchemexpress LLC HA/Hemagglutinin H1 5ug | 5ug
SDP
Catalog No. 5000186341
Encompass_Preferred
The hemagglutinin (HA) protein is essential for viral attachment to host cells binding to sialic acid receptors and initiating virion internalization through clathrin-dependent or -independent pathways As a class I viral fusion protein HA determines host range and virulence mediating viral penetration into the cytoplasm Hemagglutinin/HA Protein H17N10 (AFC35418 sf9 His) is the recombinant Virus-derived Hemagglutinin/HA protein expressed by Sf9 insect cells with C-His labeled tag
Catalog No. 50-001-86341 Supplier Medchemexpress LLC Supplier No. HYP770235UG
The hemagglutinin (HA) protein is essential for viral attachment to host cells binding to sialic acid receptors and initiating virion internalization through clathrin-dependent or -independent pathways As a class I viral fusion protein HA determines host range and virulence mediating viral penetration into the cytoplasm Hemagglutinin/HA Protein H17N10 (AFC35418 sf9 His) is the recombinant Virus-derived Hemagglutinin/HA protein expressed by Sf9 insect cells with C-His labeled tag