Learn More
Medchemexpress LLC HA/Hemagglutinin H1 5ug | 5ug
SDP
Supplier:  Medchemexpress LLC HYP770215UG
Encompass_Preferred
The hemagglutinin HA1 protein is essential for attachment of virions to host cells by binding to sialic acid-containing receptors inducing virion internalization via clathrin-dependent or clathrin- and caveolin-independent pathways As a class I viral fusion protein HA1 determines host range restriction and virulence mediating fusion between the virion membrane and the endosomal membrane Hemagglutinin/HA Protein H15N2 (ABB90704 sf9 His) is the recombinant Virus-derived Hemagglutinin/HA protein expressed by Sf9 insect cells with C-His labeled tag
Catalog No. 50-001-85950