Learn More
Medchemexpress LLC HA/Hemagglutinin H5 5ug
SDP
Catalog No. 5000179919
Encompass_Preferred
HA/Hemagglutinin Protein is a viral membrane protein responsible for the initial steps of the entry of influenza virus into the host cell It mediates binding of the virus particle to the host-cell membrane and catalyzes fusion of the viral membrane with that of the host HA is pivotal in determining virus host range and virulence HA/Hemagglutinin Protein H5N1 (AAT76166 sf9 His) is the recombinant Virus-derived HA/Hemagglutinin protein expressed by Sf9 insect cells with C-His labeled tag
Catalog No. 50-001-79919 Supplier Medchemexpress LLC Supplier No. HYP740085UG
Add to Cart
Add to Cart
HA/Hemagglutinin Protein is a viral membrane protein responsible for the initial steps of the entry of influenza virus into the host cell It mediates binding of the virus particle to the host-cell membrane and catalyzes fusion of the viral membrane with that of the host HA is pivotal in determining virus host range and virulence HA/Hemagglutinin Protein H5N1 (AAT76166 sf9 His) is the recombinant Virus-derived HA/Hemagglutinin protein expressed by Sf9 insect cells with C-His labeled tag