Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HA/Hemagglutinin H9 | >95.0% | 50 UG
SDP

Catalog No. 5000291698
Encompass_Preferred

Recombinant influenza A hemagglutinin H9N2 protein expressed in Sf9 insect cells with a His tag. Supplied as a 0.22 μm filtered solution in 20 mM Tris, 500 mM NaCl, 10% glycerol, pH 8.0. Purity is greater than 95% by reducing SDS-PAGE and endotoxin is <1 EU/μg. Recommended storage is -80°C; stable at -20°C for short-term storage. Intended for use in immunoassays, structural studies, and antibody development.

  • Recombinant protein expressed in Sf9 insect cells.
  • N-terminal His tag for purification and detection.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg).
  • Supplied in Tris/NaCl/glycerol buffer, 0.22 μm filtered.
  • Stable at -80°C; aliquot to minimize freeze-thaw cycles.
  • Suitable for ELISA, SPR (buffer exchange recommended), western blot, and antibody production.

Catalog No. 50-002-91698 Supplier Medchemexpress LLC Supplier No. HYP7501250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant influenza A hemagglutinin H9N2 protein expressed in Sf9 insect cells with a His tag. Supplied as a 0.22 μm filtered solution in 20 mM Tris, 500 mM NaCl, 10% glycerol, pH 8.0. Purity is greater than 95% by reducing SDS-PAGE and endotoxin is <1 EU/μg. Recommended storage is -80°C; stable at -20°C for short-term storage. Intended for use in immunoassays, structural studies, and antibody development.

  • Recombinant protein expressed in Sf9 insect cells.
  • N-terminal His tag for purification and detection.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg).
  • Supplied in Tris/NaCl/glycerol buffer, 0.22 μm filtered.
  • Stable at -80°C; aliquot to minimize freeze-thaw cycles.
  • Suitable for ELISA, SPR (buffer exchange recommended), western blot, and antibody production.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.