Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HA/hemagglutinin protein, H10N8 (EPI497477, sf9, His) | >90.0% | 50 UG
SDP

Catalog No. 5000293705
Encompass_Preferred

Recombinant hemagglutinin (HA) protein from Influenza A H10N8, expressed in Sf9 insect cells and purified for use in serology, antibody characterization, structural studies, and assay development. The protein comprises residues L17-D524 with a C-terminal His tag and an approximate molecular weight of 57.9 kDa.

  • Expressed in Sf9 insect cells.
  • C-terminal His tag for affinity purification and detection.
  • Sequence corresponds to residues L17-D524.
  • Approximate molecular weight 57.9 kDa.
  • Purity >90% as determined by reducing SDS-PAGE.
  • Supplied as 50 μg.

Catalog No. 50-002-93705 Supplier Medchemexpress LLC Supplier No. HYP7514150UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant hemagglutinin (HA) protein from Influenza A H10N8, expressed in Sf9 insect cells and purified for use in serology, antibody characterization, structural studies, and assay development. The protein comprises residues L17-D524 with a C-terminal His tag and an approximate molecular weight of 57.9 kDa.

  • Expressed in Sf9 insect cells.
  • C-terminal His tag for affinity purification and detection.
  • Sequence corresponds to residues L17-D524.
  • Approximate molecular weight 57.9 kDa.
  • Purity >90% as determined by reducing SDS-PAGE.
  • Supplied as 50 μg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.