Learn More
Medchemexpress LLC HA/Hemagglutinin protein, H10N8 (EPI497477, sf9, His) | 93.5% | 10 UG
SDP
Catalog No. 5000292779
Encompass_Preferred
Recombinant HA/Hemagglutinin protein (H10N8) corresponding to accession EPI497477 (L17-D524), expressed in Sf9 insect cells with a C-terminal His tag. Supplied as a 10 μg research-use reagent and intended for laboratory applications; not for human or clinical use.
  • Recombinant influenza A hemagglutinin H10N8 protein.
  • Expressed in Sf9 insect cells for proper folding.
  • C-terminal His tag for affinity purification.
  • High purity as determined by reducing SDS-PAGE (93.5%).
  • Suitable for immunology and binding assays.
  • Intended for research use only, not for human use.
Catalog No. 50-002-92779 Supplier Medchemexpress LLC Supplier No. HYP7514110UG
Recombinant HA/Hemagglutinin protein (H10N8) corresponding to accession EPI497477 (L17-D524), expressed in Sf9 insect cells with a C-terminal His tag. Supplied as a 10 μg research-use reagent and intended for laboratory applications; not for human or clinical use.
  • Recombinant influenza A hemagglutinin H10N8 protein.
  • Expressed in Sf9 insect cells for proper folding.
  • C-terminal His tag for affinity purification.
  • High purity as determined by reducing SDS-PAGE (93.5%).
  • Suitable for immunology and binding assays.
  • Intended for research use only, not for human use.