Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HA/hemagglutinin protein, H16N3 | >95.0% | 50 UG
SDP

Catalog No. 5000290551
Encompass_Preferred

Recombinant hemagglutinin (HA) protein from Influenza A H16N3 (accession AAV91217), expressed in Sf9 insect cells with a C-terminal His tag. Supplied as a 0.22 μm filtered solution in 20 mM Tris, 500 mM NaCl, 10% glycerol, pH 7.4; molecular weight ≈58.6 kDa and endotoxin <1 EU/μg. Intended for antibody generation, ELISA, and structural or functional studies. Store at -80 °C; shipped on dry ice.

  • Recombinant hemagglutinin from Influenza A H16N3.
  • Expressed in Sf9 insect cells to support proper folding.
  • C-terminal His tag to facilitate purification and detection.
  • Supplied as a 0.22 μm filtered formulation for stability.
  • Formulation: 20 mM Tris, 500 mM NaCl, 10% glycerol, pH 7.4.
  • Endotoxin level below 1 EU per microgram.
  • Suitable for immunization, ELISA, and biophysical characterization.

Catalog No. 50-002-90551 Supplier Medchemexpress LLC Supplier No. HYP7644550UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant hemagglutinin (HA) protein from Influenza A H16N3 (accession AAV91217), expressed in Sf9 insect cells with a C-terminal His tag. Supplied as a 0.22 μm filtered solution in 20 mM Tris, 500 mM NaCl, 10% glycerol, pH 7.4; molecular weight ≈58.6 kDa and endotoxin <1 EU/μg. Intended for antibody generation, ELISA, and structural or functional studies. Store at -80 °C; shipped on dry ice.

  • Recombinant hemagglutinin from Influenza A H16N3.
  • Expressed in Sf9 insect cells to support proper folding.
  • C-terminal His tag to facilitate purification and detection.
  • Supplied as a 0.22 μm filtered formulation for stability.
  • Formulation: 20 mM Tris, 500 mM NaCl, 10% glycerol, pH 7.4.
  • Endotoxin level below 1 EU per microgram.
  • Suitable for immunization, ELISA, and biophysical characterization.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.