Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HA/hemagglutinin protein, H16N3 (AAV91217, sf9, His) | 97.1%
SDP

Catalog No. 5000290344
Encompass_Preferred

Recombinant hemagglutinin (HA) protein from influenza A H16N3, produced in Sf9 insect cells and supplied with a C-terminal His tag. The protein is provided for research use and is suitable for studies of viral attachment, host range, and immunological assays. Manufacturer documentation includes a product datasheet and certificate of analysis confirming molecular weight and purity.

  • Recombinant H16N3 hemagglutinin suitable for binding and immunoassays.
  • Expressed in Sf9 insect cells for eukaryotic processing.
  • C-terminal His tag for affinity purification and detection.
  • Approximately 58.6 kDa molecular weight.
  • Purity 97.1% as reported on the certificate of analysis.
  • Product datasheet and COA available for quality verification.

Catalog No. 50-002-90344 Supplier Medchemexpress LLC Supplier No. HYP7644510UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant hemagglutinin (HA) protein from influenza A H16N3, produced in Sf9 insect cells and supplied with a C-terminal His tag. The protein is provided for research use and is suitable for studies of viral attachment, host range, and immunological assays. Manufacturer documentation includes a product datasheet and certificate of analysis confirming molecular weight and purity.

  • Recombinant H16N3 hemagglutinin suitable for binding and immunoassays.
  • Expressed in Sf9 insect cells for eukaryotic processing.
  • C-terminal His tag for affinity purification and detection.
  • Approximately 58.6 kDa molecular weight.
  • Purity 97.1% as reported on the certificate of analysis.
  • Product datasheet and COA available for quality verification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.