Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HA/hemagglutinin protein, H18N11 (AGX84934, sf9, His) | >90.0% | 10 UG
SDP

Catalog No. 5000291414
Encompass_Preferred

Recombinant hemagglutinin H18N11 protein expressed in Sf9 insect cells with a C-terminal His tag. Supplied as a filtered, buffered solution formulated for research use in immunological assays, antibody development, and biochemical characterization. Store frozen and avoid repeated freeze-thaw cycles.

  • C-terminal His tag for purification and detection.
  • Expressed in Sf9 insect cells for eukaryotic folding.
  • Supplied as a 0.22 μm filtered solution in 20 mM Tris, 500 mM NaCl, 10% glycerol, pH 7.4.
  • Protein sequence range D15-D513 for domain-specific studies.
  • Greater than 90.0% purity as determined by reducing SDS-PAGE.
  • Available in small research sizes suitable for pilot experiments.
  • Store at -80°C for long term; stable at -20°C short term; avoid repeated freeze-thaw.

Catalog No. 50-002-91414 Supplier Medchemexpress LLC Supplier No. HYP7513510UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant hemagglutinin H18N11 protein expressed in Sf9 insect cells with a C-terminal His tag. Supplied as a filtered, buffered solution formulated for research use in immunological assays, antibody development, and biochemical characterization. Store frozen and avoid repeated freeze-thaw cycles.

  • C-terminal His tag for purification and detection.
  • Expressed in Sf9 insect cells for eukaryotic folding.
  • Supplied as a 0.22 μm filtered solution in 20 mM Tris, 500 mM NaCl, 10% glycerol, pH 7.4.
  • Protein sequence range D15-D513 for domain-specific studies.
  • Greater than 90.0% purity as determined by reducing SDS-PAGE.
  • Available in small research sizes suitable for pilot experiments.
  • Store at -80°C for long term; stable at -20°C short term; avoid repeated freeze-thaw.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.