Learn More
Medchemexpress LLC HA/hemagglutinin protein, H18N11 (AGX84934, sf9, His) | >90.0% | 5 UG
SDP
Catalog No. 5000293154
Encompass_Preferred
HA/Hemagglutinin protein H18N11 is a recombinant Influenza A hemagglutinin fragment (residues D15-D513) expressed in Sf9 insect cells with a C-terminal His tag. Supplied in research-scale vials (including 5 μg), the protein is intended for use in binding assays, antibody generation, and structural or functional studies. Store at -80°C and ship on dry ice.
Catalog No. 50-002-93154 Supplier Medchemexpress LLC Supplier No. HYP751355UG
HA/Hemagglutinin protein H18N11 is a recombinant Influenza A hemagglutinin fragment (residues D15-D513) expressed in Sf9 insect cells with a C-terminal His tag. Supplied in research-scale vials (including 5 μg), the protein is intended for use in binding assays, antibody generation, and structural or functional studies. Store at -80°C and ship on dry ice.