Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HA/hemagglutinin protein, H1N1 | >95.0% | 10 UG
SDP

Catalog No. 5000290051
Encompass_Preferred

Recombinant HA (hemagglutinin) protein from Influenza A H1N1 (A/California/04/2009), expressed in Sf9 insect cells and supplied as a tag-free, lyophilized full-length mature protein for research use in antigenicity studies, antibody development, and assay validation.

  • Tag-free, full-length mature HA protein.
  • Expressed in Sf9 insect cells.
  • Purity >95.0% by reducing SDS-PAGE.
  • Lyophilized from 20 mM Tris, 150 mM NaCl, 10% glycerol, pH 7.5.
  • Molecular weight approximately 61.5 kDa.
  • Reconstitution and storage guidance provided; avoid reconstituting below 100 μg/mL.

Catalog No. 50-002-90051 Supplier Medchemexpress LLC Supplier No. HYP7644710UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant HA (hemagglutinin) protein from Influenza A H1N1 (A/California/04/2009), expressed in Sf9 insect cells and supplied as a tag-free, lyophilized full-length mature protein for research use in antigenicity studies, antibody development, and assay validation.

  • Tag-free, full-length mature HA protein.
  • Expressed in Sf9 insect cells.
  • Purity >95.0% by reducing SDS-PAGE.
  • Lyophilized from 20 mM Tris, 150 mM NaCl, 10% glycerol, pH 7.5.
  • Molecular weight approximately 61.5 kDa.
  • Reconstitution and storage guidance provided; avoid reconstituting below 100 μg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.