Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ha/hemagglutinin protein, h1n1 | >95.0% | 50 UG
SDP

Catalog No. 5000290497
Encompass_Preferred

Recombinant HA (hemagglutinin) H1N1 protein expressed in Sf9 insect cells, supplied as a lyophilized, tag-free preparation. Designed for ELISA, antibody validation, receptor-binding, and structural studies, the product offers high purity and low endotoxin for sensitive biochemical and immunological assays.

  • Recombinant hemagglutinin H1N1 protein expressed in Sf9 insect cells.
  • Lyophilized, tag-free format for flexibility in assay setup.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Endotoxin below 1 EU/μg supporting low-background assays.
  • Formulated in Tris, NaCl, and glycerol for stability.
  • Stable frozen; reconstitute to ≥100 μg/mL and aliquot for storage.
  • Suitable for ELISA, antibody generation, and receptor-binding studies.

Catalog No. 50-002-90497 Supplier Medchemexpress LLC Supplier No. HYP7644750UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant HA (hemagglutinin) H1N1 protein expressed in Sf9 insect cells, supplied as a lyophilized, tag-free preparation. Designed for ELISA, antibody validation, receptor-binding, and structural studies, the product offers high purity and low endotoxin for sensitive biochemical and immunological assays.

  • Recombinant hemagglutinin H1N1 protein expressed in Sf9 insect cells.
  • Lyophilized, tag-free format for flexibility in assay setup.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Endotoxin below 1 EU/μg supporting low-background assays.
  • Formulated in Tris, NaCl, and glycerol for stability.
  • Stable frozen; reconstitute to ≥100 μg/mL and aliquot for storage.
  • Suitable for ELISA, antibody generation, and receptor-binding studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.