Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HA/hemagglutinin protein, H1N1 (ACP41105, sf9) | >95.0% | 100 UG
SDP

Catalog No. 5000290380
Encompass_Preferred

Recombinant hemagglutinin (HA) protein from Influenza A H1N1 (A/California/04/2009), expressed in Sf9 insect cells and supplied lyophilized. The product is intended for research applications including serological assays, antibody generation, and studies of viral entry, offering high purity and defined formulation for consistent results.

  • Recombinant full-length HA protein representative of H1N1 strain.
  • Expressed in Sf9 insect cells to support eukaryotic post-translational modifications.
  • Lyophilized formulation for extended shelf life and convenient storage.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Formulated in Tris-buffered saline with glycerol for stability.
  • Suitable for serological assays, antibody production, and viral entry studies.
  • Store frozen; after reconstitution, stable at 4°C short term or -20°C for longer storage.

Catalog No. 50-002-90380 Supplier Medchemexpress LLC Supplier No. HYP76447100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant hemagglutinin (HA) protein from Influenza A H1N1 (A/California/04/2009), expressed in Sf9 insect cells and supplied lyophilized. The product is intended for research applications including serological assays, antibody generation, and studies of viral entry, offering high purity and defined formulation for consistent results.

  • Recombinant full-length HA protein representative of H1N1 strain.
  • Expressed in Sf9 insect cells to support eukaryotic post-translational modifications.
  • Lyophilized formulation for extended shelf life and convenient storage.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Formulated in Tris-buffered saline with glycerol for stability.
  • Suitable for serological assays, antibody production, and viral entry studies.
  • Store frozen; after reconstitution, stable at 4°C short term or -20°C for longer storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.