Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HA/hemagglutinin protein, H5N6 (AKS48023, Sf9, His) | >95.0% | 50 UG
SDP

Catalog No. 5000293689
Encompass_Preferred

Recombinant hemagglutinin (HA) protein derived from Influenza A H5N6 (A/yunnan/0127/2015). The protein is produced in Sf9 insect cells with a His affinity tag and is intended as a purified reagent for research applications such as immunoassays, antibody generation, and biochemical studies.

  • Recombinant HA corresponding to Influenza A H5N6.
  • Expressed in Sf9 insect cells.
  • His-tagged for affinity purification.
  • Purity >95.0% as determined by reducing SDS-PAGE.
  • Supplied as a 50 μg format.
  • Suitable for ELISA, western blot, and antibody production.
  • Stable when stored frozen per product documentation.

Catalog No. 50-002-93689 Supplier Medchemexpress LLC Supplier No. HYP7502750UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant hemagglutinin (HA) protein derived from Influenza A H5N6 (A/yunnan/0127/2015). The protein is produced in Sf9 insect cells with a His affinity tag and is intended as a purified reagent for research applications such as immunoassays, antibody generation, and biochemical studies.

  • Recombinant HA corresponding to Influenza A H5N6.
  • Expressed in Sf9 insect cells.
  • His-tagged for affinity purification.
  • Purity >95.0% as determined by reducing SDS-PAGE.
  • Supplied as a 50 μg format.
  • Suitable for ELISA, western blot, and antibody production.
  • Stable when stored frozen per product documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.