Learn More
Medchemexpress LLC HY-P0239 10mg , HA Peptide CAS:92000-76-5 Purity:>98%
SDP
Catalog No. 502259841
Encompass_Preferred
HY-P0239 10mg HA Peptide CAS: 92000-76-5 HA Peptide (HA tag) is a nine amino acids peptide derived from the human influenza hemagglutinin (HA). HA Peptide is extensively used to isolate, purify, detect, and track the protein of interest in cell biology and biochemistry. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9841 Supplier Medchemexpress LLC Supplier No. HYP023910MG
HY-P0239 10mg HA Peptide CAS: 92000-76-5 HA Peptide (HA tag) is a nine amino acids peptide derived from the human influenza hemagglutinin (HA). HA Peptide is extensively used to isolate, purify, detect, and track the protein of interest in cell biology and biochemistry. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.