Learn More
Medchemexpress LLC HY-P0239 1mg Medchemexpress, HA Peptide CAS:92000-76-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP02391MG
Encompass_Preferred
Medchemexpress, HY-P0239 1mg HA Peptide CAS:92000-76-5 HA Peptide (HA tag) is a nine amino acids peptide derived from the human influenza hemagglutinin (HA). It is extensively used to isolate, purify, detect, and track the protein of interest in cell biology and biochemistry. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4070