Learn More
Medchemexpress LLC HY-P0239A 5mg , HA Peptide (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0239A5MG
Encompass_Preferred
HY-P0239A 5mg HA Peptide (TFA) CAS: HA Peptide (TFA) is a nine amino acids peptide derived from the human influenza hemagglutinin (HA). HA Peptide (TFA) is extensively used to isolate, purify, detect, and track the protein of interest in cell biology and biochemistry. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1566