Learn More
Enzo Life Sciences HA-tag polyclonal antibody (biotin conjugate) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2900100
Encompass_Preferred
Isotype: IgG. Immunogen: Synthetic peptide corresponding to YPYDVPDYA of the HA tag. Species reactivity: Species independent. Applications: WB. Purity: Antigen affinity chromatography. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.05% sodium azide.
Catalog No. 50-201-2830