Learn More
AdipoGen HA-Ubiquitin-vinyl sulfone (human) (rec.) (HA) (50 µg)
SDP
Supplier:  AdipoGen SBBPS0122C050
Encompass_Preferred
Protein. Human ubiquitin (aa1-76) (Accession Nr. P0CG47) with a C-terminal vinyl sulfone group and a N-terminal HA-Tag. Source: E. coli. Liquid. In 50mM MES pH 6.0, 100mM sodium chloride. Purity: >97% (LCMS). HA-Ubiquitin vinyl sulfone is a potent, irreversible and specific inhibitor of deubiquitinating enzymes (DUBs) based on a C-terminal electrophilic vinyl sulfone group. It targets four of the five major DUB families: UCH (Ubiquitin c-terminal hydrolases), USP (Ubiquitin Specific Proteases), OTU (Ovarian Tumor Proteases), and MJD (Machado-Josephin domain proteases) while JAMM metalloproteases are not inhibited. HA-Ubiquitin vinyl sulfone can be used for activity profiling experiments and using the HA-Tag conveniently for Immunoprecipitations in search for new or novel enzymes reacting with the c-terminal warhead.
Catalog No. 50-205-4703