Learn More
Enzo Life Sciences Ubiquitin vinyl sulfone, (HA-tag) (25 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW01550025
Encompass_Preferred
For use in the detection and identification of deubiquitinylating enzymes. HA-Ub-VS is a DUB active site directed probe generated by a chemical ligation method, that acts as a potent and irreversible inhibitor of DUBs through covalent modification of the active site and as a specific probe for enzymes with DUB activity. The HA peptide sequence (YPYDVPDYA), derived from the influenza hemagglutinin protein, facilitates sensitive identification or purification of HA-Ub-VS modified DUBs through recognition by anti-HA antibodies and/or anti-HA-agarose. For use in the detection and identification of deubiquitinylating enzymes. HA-Ub-VS is a DUB active site directed probe generated by a chemical ligation method, that acts as a potent and irreversible inhibitor of DUBs through covalent modification of the active site and as a specific probe for enzymes with DUB activity. Alternative name: HA-Ub-VS. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized. Source: Synthetic.
Catalog No. 50-201-2232