Learn More
Cayman Chemical HA-1077 (hydrochloride), Purity: ≥98%, CAS: 203911-27-7, 50 mg
SDP
Catalog No. 502889644
Encompass_Preferred
A potent inhibitor of ROCK2, PRK-2, MSK1, and MAPKAP-K1b with IC50 values of 1.9, 4, 5, and 15 µM, respectively; has been shown to reduce blood vessel constriction, decreases pulmonary arterial pressure, inhibit tumor angiogenesis, and improve insulin signaling in a diabetic rat model; drug forms are marketed for the treatment of pulmonary arterial hypertension and stable angina
Catalog No. 50-288-9644 Supplier Cayman Chemical Supplier No. 1001055950 MG
A potent inhibitor of ROCK2, PRK-2, MSK1, and MAPKAP-K1b with IC50 values of 1.9, 4, 5, and 15 µM, respectively; has been shown to reduce blood vessel constriction, decreases pulmonary arterial pressure, inhibit tumor angiogenesis, and improve insulin signaling in a diabetic rat model; drug forms are marketed for the treatment of pulmonary arterial hypertension and stable angina