Learn More
Medchemexpress LLC HY-19329 5mg Medchemexpress, HA130 CAS:1229652-21-4 Purity:>98%
SDP
Catalog No. 502030847
Encompass_Preferred
Medchemexpress, HY-19329 5mg HA130 CAS:1229652-21-4 HA130 is a selective autotaxin (ATX) inhibitor with an IC50 of 28 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0847 Supplier Medchemexpress LLC Supplier No. HY193295MG
Medchemexpress, HY-19329 5mg HA130 CAS:1229652-21-4 HA130 is a selective autotaxin (ATX) inhibitor with an IC50 of 28 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.