Learn More
Medchemexpress LLC HY-12011 10mg Medchemexpress, HA14-1 CAS:65673-63-4 Purity:>98%
SDP
Catalog No. 501969577
Encompass_Preferred
Medchemexpress, HY-12011 10mg HA14-1 CAS:65673-63-4 HA14-1 is a Bcl-2/Bcl-XL antagonist. HA14-1 binds the designated pocket on Bcl-2 with the IC50 of ≈9 μM in competing with the Bcl-2 binding of Flu-BakBH3, and inhibits its function. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9577 Supplier Medchemexpress LLC Supplier No. HY1201110MG
Medchemexpress, HY-12011 10mg HA14-1 CAS:65673-63-4 HA14-1 is a Bcl-2/Bcl-XL antagonist. HA14-1 binds the designated pocket on Bcl-2 with the IC50 of ≈9 μM in competing with the Bcl-2 binding of Flu-BakBH3, and inhibits its function. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.