Learn More
Abcam HA14-1, Bcl-2 inhibitor. BH-3 mimetic., 50MG
Catalog No. 502706697
Encompass_Preferred
MW 409.2 Da. Cell permeable Bcl-2 inhibitor. BH-3 mimetic. Disrupts Bax and Bcl-2 interactions. Apoptotic anticancer agent.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6697 Supplier Abcam Supplier No. AB14130650MG
MW 409.2 Da. Cell permeable Bcl-2 inhibitor. BH-3 mimetic. Disrupts Bax and Bcl-2 interactions. Apoptotic anticancer agent.
The product is subject to the following: Abcam Restricted Use Statement