Learn More
Medchemexpress LLC HY-116100A 5mg Medchemexpress, HA155 CAS:1312201-00-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY116100A5MG
Encompass_Preferred
Medchemexpress, HY-116100A 5mg HA155 CAS:1312201-00-5 HA-155 is a potent and selective autotaxin (ATX) inhibitor with an IC50 of 5.7 nM[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9406