Learn More
Thermo Scientific™ HAAKE™ Viscotester™ iQ Rheometer Application Packages
Description
Ready to use in seconds in any configuration you need
Acquire simple viscosity curves or perform complex rheological investigations covering measurements of low viscous liquids up to highly viscous pastes with the Thermo Scientific™ HAAKE™ Viscotester™ rheometer, which can be used as a standalone unit or optionally, be controlled by software. The size and footprint of the HAAKE™ Viscotester™ rheometer make it a portable unit suitable for mobile use or allow it to serve as an important tool in the quality control laboratory.
Application packages are designed to perform a diverse set of rheological tasks and meet a variety of application needs.
- Advanced design with one-hand lift operation
- Touchscreen control panel with multilingual, graphical user interface
- Data quality assessment and QC quality criteria
- Connect Assist quick coupling and automatic recognition of temperature modules and rotors
- Liquid temperature module uses a circulator for coaxial cylinders and parallel plates
- Self-sufficient Peltier temperature module for coaxial cylinders and parallel plates
- Small dimensioned, coaxial cylinders use less sample and shorter temperature equilibration times to minimize costs per measurement
- Universal container holder for relative measurements in the original containers
Specifications
Specifications
| Product Type | Rheometer Application Package |
| Includes | HAAKE Viscotester iQ Rheometer, Liquid Temperature Module TM-LI-C32, Cup CCB25 DIN and Rotor CCB25 DIN/Ti |
| For Use With (Application) | Liquids |