Learn More
Sigma Aldrich Fine Chemicals Biosciences HABA/Avidin Reagent lyophi
Supplier:  Sigma Aldrich Fine Chemicals Biosciences H21531VL
Encompass
HABA/Avidin Reagent is specific for chromogenic applications that depend upon quantification of biotin.
Catalog No. 50-290-1467