Learn More
ACROBiosystems Recombinant Protein;Human HABP1 / C1QBP Protein;E.coli;1MG;C1P,C1QBP,GC1QBP,HABP1,SF2P32,p33
Catalog No. 501658897
Encompass_Preferred
Recombinant Protein;Human HABP1 / C1QBP Protein;1MG;Human HABP1, His Tag (C1P-H5122) is expressed from E.coli cells. It contains AA His 75 - Gln 282 (Accession # AAH00435).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.6 kDa. The protein migrates as 35 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8897 Supplier ACROBiosystems Supplier No. C1PH51221MG
Recombinant Protein;Human HABP1 / C1QBP Protein;1MG;Human HABP1, His Tag (C1P-H5122) is expressed from E.coli cells. It contains AA His 75 - Gln 282 (Accession # AAH00435).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.6 kDa. The protein migrates as 35 kDa under reducing (R) condition (SDS-PAGE).