Learn More
Aviva Systems Biology Corp Habp2 Antibody - C-terminal region (ARP54719_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54719_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Habp2. It was validated on Western Blot. . Alternate names: AI035669, FSAP, HABP, HGFAL, MGC28705, PHBPApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-960