Learn More
Aviva Systems Biology Corp HABP2 Antibody - middle region (ARP54718_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54718_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HABP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HABP2. Alternate names: FSAP, HABP, HGFAL, PHBPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61291