Learn More
ATCC HAC15; Adrenal Carcinoma; Human (Homo sapiens)
Catalog No. 502382525
Encompass
HAC15 is an epithelial-like cell line that was clonally isolated from the adrenal gland of a 48-year-old, Black female patient with carcinoma. HAC15 provides an important model system for defining the molecular mechanisms regulating aldosterone and cortisol production and can be applied to studies of either normal adrenal cell function or adrenocortical cancer. This cell line was deposited by WE Rainey.
Catalog No. 50-238-2525 Supplier ATCC Supplier No. CRL3301
HAC15 is an epithelial-like cell line that was clonally isolated from the adrenal gland of a 48-year-old, Black female patient with carcinoma. HAC15 provides an important model system for defining the molecular mechanisms regulating aldosterone and cortisol production and can be applied to studies of either normal adrenal cell function or adrenocortical cancer. This cell line was deposited by WE Rainey.