Learn More
ADDEXBIO TECHNOLOGIES HACAT CELLS (T0020001
Catalog No. NC0309203
Encompass
In vitro spontaneously transformed keratinocytes from histologically normal skin. Species: Homo sapiens. Keratinocyte. Adherent monolayer. Patient: Male, Caucasian, 62 years of age. Medium: AddexBio-Optimized DMEM (C0003-02) + 10% FBS. Subculture: Recommended 1:5 to 1:10 using 0.25% trypsin or trypsin/EDTA, 5% CO2; 37 degrees Celsius. Bacteria: Negative. Yeast: Negative. Mycoplasma: Negative. HIV: Negative. Hepatitis B: Negative. Hepatitis C: Negative.
Catalog No. NC0309203 Supplier ADDEXBIO TECHNOLOGIES Supplier No. T0020001
In vitro spontaneously transformed keratinocytes from histologically normal skin. Species: Homo sapiens. Keratinocyte. Adherent monolayer. Patient: Male, Caucasian, 62 years of age. Medium: AddexBio-Optimized DMEM (C0003-02) + 10% FBS. Subculture: Recommended 1:5 to 1:10 using 0.25% trypsin or trypsin/EDTA, 5% CO2; 37 degrees Celsius. Bacteria: Negative. Yeast: Negative. Mycoplasma: Negative. HIV: Negative. Hepatitis B: Negative. Hepatitis C: Negative.