Learn More
Hach Company Hach 3/4 in Combination pH/ORP Sensor, Convertible Style, PPS body, HF-resistant glass, 15 ft cable
Catalog No. 5000207854 Shop All Hach Company Products
Encompass
The Hach 3/4 Inch Combination pH/ORP Sensor (PC1R3A) is a convertible-style immersion or in-line sensor designed for reliable pH measurement in applications that require HF resistance. It features a durable PPS body, HF-resistant glass electrode, and a fixed 15 ft (4.5 m) cable with stripped and tinned leads.
  • Convertible mounting for immersion or in-line use.
  • HF-resistant glass pH electrode for aggressive environments.
  • PPS body for chemical and mechanical resistance.
  • Fixed 15 ft (4.5 m) cable terminated with stripped and tinned wires.
  • Built-in PT1000 temperature compensation.
  • Measurement range 0-14 pH with ±0.1 pH accuracy.
  • Operating temperature 0-105 °C and pressure rating 6.9 bar at 100 °C.
  • 0.75 in NPT sensor thread for standard process fittings.
Catalog No. 50-002-07854 Supplier Hach Company Supplier No. PC1R3A
The Hach 3/4 Inch Combination pH/ORP Sensor (PC1R3A) is a convertible-style immersion or in-line sensor designed for reliable pH measurement in applications that require HF resistance. It features a durable PPS body, HF-resistant glass electrode, and a fixed 15 ft (4.5 m) cable with stripped and tinned leads.
  • Convertible mounting for immersion or in-line use.
  • HF-resistant glass pH electrode for aggressive environments.
  • PPS body for chemical and mechanical resistance.
  • Fixed 15 ft (4.5 m) cable terminated with stripped and tinned wires.
  • Built-in PT1000 temperature compensation.
  • Measurement range 0-14 pH with ±0.1 pH accuracy.
  • Operating temperature 0-105 °C and pressure rating 6.9 bar at 100 °C.
  • 0.75 in NPT sensor thread for standard process fittings.