Learn More
Hach Company Hach Acid Solution for EZ4003 and EZ4004 Total Alkalinity Analyzer, 0.025 N HCl, 2 L
Catalog No. NC3839294 Shop All Hach Company Products
Encompass
Acid solution formulated for use with EZ Series online total alkalinity analyzers. This ready-to-use reagent contains 0.025 N hydrochloric acid for titration in the 10-250 mg/L alkalinity measurement range and is supplied in a 2 L pack designed for direct use in EZ Series reagent systems.
  • Ready-to-use 0.025 N hydrochloric acid reagent.
  • Titration range suitable for 10-250 mg/L alkalinity measurements.
  • Supplied in a 2 L volume for analyzer reagent reservoirs.
  • Prepared for direct use with EZ Series online alkalinity analyzers.
  • Packaged for laboratory and field reagent replacement workflows.
Catalog No. NC3839294 Supplier Hach Company Supplier No. APPC400401
Acid solution formulated for use with EZ Series online total alkalinity analyzers. This ready-to-use reagent contains 0.025 N hydrochloric acid for titration in the 10-250 mg/L alkalinity measurement range and is supplied in a 2 L pack designed for direct use in EZ Series reagent systems.
  • Ready-to-use 0.025 N hydrochloric acid reagent.
  • Titration range suitable for 10-250 mg/L alkalinity measurements.
  • Supplied in a 2 L volume for analyzer reagent reservoirs.
  • Prepared for direct use with EZ Series online alkalinity analyzers.
  • Packaged for laboratory and field reagent replacement workflows.