Learn More
Hach Company Hach BioTector TN Cleaning Solution, 18.9 L - for BioTector/B7000 TN analyzers
Catalog No. NC3821305 Shop All Hach Company Products
Encompass
Hach BioTector TN Cleaning Solution is a pre-formulated reagent supplied in an 18.9 L plastic bottle for routine cleaning and maintenance of BioTector TN/TP online analyzers, including the B7000 series. It is intended to remove residues that can affect measurements and help maintain analyzer performance and accuracy.
  • Comes in an 18.9 L plastic bottle.
  • Formulated for cleaning TN/TP online analyzers.
  • Helps remove residues and maintain measurement accuracy.
  • Keep tightly closed in a dry, cool, well-ventilated place.
  • Considered hazardous for shipping; consult the safety data sheet before transport.
Catalog No. NC3821305 Supplier Hach Company Supplier No. 2985662
Hach BioTector TN Cleaning Solution is a pre-formulated reagent supplied in an 18.9 L plastic bottle for routine cleaning and maintenance of BioTector TN/TP online analyzers, including the B7000 series. It is intended to remove residues that can affect measurements and help maintain analyzer performance and accuracy.
  • Comes in an 18.9 L plastic bottle.
  • Formulated for cleaning TN/TP online analyzers.
  • Helps remove residues and maintain measurement accuracy.
  • Keep tightly closed in a dry, cool, well-ventilated place.
  • Considered hazardous for shipping; consult the safety data sheet before transport.