Learn More
Hach Company Hach Flow Cell for 1 in NPT Sensor, Model 7300800 - Fits pHD, NH4D, NO3D Sensors
Catalog No. 5000207225 Shop All Hach Company Products
Encompass
Inline flow cell designed to house 1 in NPT stainless-steel sensors for pH and ion-selective measurements. The clear-body design enables visual inspection of the sensor and accumulated debris, and tangential inlet porting creates a vortex to separate heavy particles and improve mixing, simplifying maintenance and ensuring reliable sample flow.
  • Clear body enables quick visual inspection of sensor and debris.
  • Tangential inlet porting separates heavy particles and improves mixing.
  • 3/8 in inlet and 1/2 in outlet for common process connections.
  • Designed for 1 in NPT sensor mounting in inline systems.
  • See-through housing simplifies maintenance and troubleshooting.
Catalog No. 50-002-07225 Supplier Hach Company Supplier No. 7300800
Add to Cart
Add to Cart
Inline flow cell designed to house 1 in NPT stainless-steel sensors for pH and ion-selective measurements. The clear-body design enables visual inspection of the sensor and accumulated debris, and tangential inlet porting creates a vortex to separate heavy particles and improve mixing, simplifying maintenance and ensuring reliable sample flow.
  • Clear body enables quick visual inspection of sensor and debris.
  • Tangential inlet porting separates heavy particles and improves mixing.
  • 3/8 in inlet and 1/2 in outlet for common process connections.
  • Designed for 1 in NPT sensor mounting in inline systems.
  • See-through housing simplifies maintenance and troubleshooting.