Learn More
Hach Company Hach Graduated Cylinder, Polymethylpentene (PMP), 25 mL, 0.5 mL Divisions, ±0.3 mL (Class B), Raised Graduations, Shatter-Resistant
Catalog No. 5000207334 Shop All Hach Company Products
Encompass
This 25 mL polymethylpentene (PMP) graduated cylinder provides accurate, repeatable volume measurements for general laboratory use. It is transparent, shatter-resistant, and chemically inert, with raised graduations that minimize meniscus reading error. Calibrated to contain and to deliver at 20 °C, it offers Class B accuracy for routine measurement tasks.
  • Shatter-resistant polymethylpentene construction.
  • Front-to-back raised graduations for easy reading.
  • 0.5 mL divisions with ±0.3 mL accuracy (Class B).
  • Calibrated to contain and to deliver at 20 °C.
  • Thermally resistant up to 135 °C (275 °F).
  • Excellent chemical resistance and biologically inert material.
  • Single metric scale; supplied as one each.
Catalog No. 50-002-07334 Supplier Hach Company Supplier No. 217240
This 25 mL polymethylpentene (PMP) graduated cylinder provides accurate, repeatable volume measurements for general laboratory use. It is transparent, shatter-resistant, and chemically inert, with raised graduations that minimize meniscus reading error. Calibrated to contain and to deliver at 20 °C, it offers Class B accuracy for routine measurement tasks.
  • Shatter-resistant polymethylpentene construction.
  • Front-to-back raised graduations for easy reading.
  • 0.5 mL divisions with ±0.3 mL accuracy (Class B).
  • Calibrated to contain and to deliver at 20 °C.
  • Thermally resistant up to 135 °C (275 °F).
  • Excellent chemical resistance and biologically inert material.
  • Single metric scale; supplied as one each.