Learn More
Hach Company Hach IntelliCAL PHC729 (PHC72901AP) Refillable RedRod Glass pH Electrode Kit with Reagents - For Hach HQ Series Laboratory and Portable Meters, 1 m Cable
Catalog No. 5000207501 Shop All Hach Company Products
Encompass
Refillable RedRod glass pH electrode kit for laboratory surface and shallow-sample measurements. The digital combination electrode includes a 1 m cable and a reagents pack with calibration buffers, reference electrolyte, cleaning and storage solutions, a storage soaker bottle, and documentation to support calibration and maintenance. Intended for use with HQ series laboratory and portable meters.
Catalog No. 50-002-07501 Supplier Hach Company Supplier No. PHC72901AP
Refillable RedRod glass pH electrode kit for laboratory surface and shallow-sample measurements. The digital combination electrode includes a 1 m cable and a reagents pack with calibration buffers, reference electrolyte, cleaning and storage solutions, a storage soaker bottle, and documentation to support calibration and maintenance. Intended for use with HQ series laboratory and portable meters.