Learn More
Hach Company Hach Kit, 1 Polyethylene 2.5 Gallon Bottle for SD900 Refrigerated Sampler (RF010030)
Catalog No. 5000207281 Shop All Hach Company Products
Encompass
Hach RF010030 is a replacement kit that provides a 2.5 gallon polyethylene sample bottle and the associated fittings needed for refrigerated sampler maintenance. The kit supplies a durable bottle with cap and a full-bottle shut-off to simplify sample collection, reduce spills, and streamline routine sampler upkeep.
  • 2.5 gallon polyethylene bottle for reliable sample storage.
  • Includes cap and full-bottle shut-off to prevent overflow.
  • Supplied tubing and tubing support for straightforward installation.
  • Suitable for refrigerated sampler applications.
  • One-bottle kit for easy replacement and maintenance.
This item is not returnable. View return policy
Catalog No. 50-002-07281 Supplier Hach Company Supplier No. RF010030
Hach RF010030 is a replacement kit that provides a 2.5 gallon polyethylene sample bottle and the associated fittings needed for refrigerated sampler maintenance. The kit supplies a durable bottle with cap and a full-bottle shut-off to simplify sample collection, reduce spills, and streamline routine sampler upkeep.
  • 2.5 gallon polyethylene bottle for reliable sample storage.
  • Includes cap and full-bottle shut-off to prevent overflow.
  • Supplied tubing and tubing support for straightforward installation.
  • Suitable for refrigerated sampler applications.
  • One-bottle kit for easy replacement and maintenance.