Learn More
Hach Company Hach Kit - 24 x 575 mL Polyethylene Bottles with Caps, Retainer and Distributor Arm for SD900 Portable Compact Sampler (PC240575)
Catalog No. 5000207509 Shop All Hach Company Products
Encompass
This kit includes 24 polyethylene sample bottles (575 mL each) plus caps, a retainer, and a distributor arm for use with SD900 portable compact samplers. It is intended as a replacement and resupply kit to support discrete field sampling and simplify maintenance of compact samplers.
  • Contains 24 polyethylene bottles, 575 mL capacity each.
  • Includes caps to secure and protect collected samples.
  • Includes a retainer and distributor arm for proper bottle alignment.
  • Compatible with SD900 portable compact samplers.
  • Designed for convenient field replacement and resupply.
Catalog No. 50-002-07509 Supplier Hach Company Supplier No. PC240575
This kit includes 24 polyethylene sample bottles (575 mL each) plus caps, a retainer, and a distributor arm for use with SD900 portable compact samplers. It is intended as a replacement and resupply kit to support discrete field sampling and simplify maintenance of compact samplers.
  • Contains 24 polyethylene bottles, 575 mL capacity each.
  • Includes caps to secure and protect collected samples.
  • Includes a retainer and distributor arm for proper bottle alignment.
  • Compatible with SD900 portable compact samplers.
  • Designed for convenient field replacement and resupply.