Learn More
Hach Company Hach MP-4 Portable Meter (HMP4) - Probeless conductivity, TDS, resistivity, and temperature meter
Catalog No. NC3834656 Shop All Hach Company Products
Encompass
The MP-4 Portable Meter is a cableless, single-handed instrument for rapid testing of conductivity, resistivity, total dissolved solids (TDS), and temperature in clean water applications. Integrated sample cups and one-button operation enable fast, probeless measurements suitable for pool and spa monitoring, water treatment, and field testing.
  • Probeless operation with integrated sample cups.
  • Measures conductivity, resistivity, TDS, and temperature.
  • Temperature accuracy of ±0.1 °C.
  • Battery life greater than 100 hours (≈5,000 tests) with auto-shutoff.
  • One-button calibration; typical calibration interval two weeks to monthly.
  • Automatic temperature compensation; adjustable or disableable.
  • User-adjustable conductivity/TDS conversion ratio for nonstandard solutions.
  • Data retained in non-volatile memory; waterproof and buoyant-operational after immersion up to 1 m (3 ft).
Catalog No. NC3834656 Supplier Hach Company Supplier No. HMP4
The MP-4 Portable Meter is a cableless, single-handed instrument for rapid testing of conductivity, resistivity, total dissolved solids (TDS), and temperature in clean water applications. Integrated sample cups and one-button operation enable fast, probeless measurements suitable for pool and spa monitoring, water treatment, and field testing.
  • Probeless operation with integrated sample cups.
  • Measures conductivity, resistivity, TDS, and temperature.
  • Temperature accuracy of ±0.1 °C.
  • Battery life greater than 100 hours (≈5,000 tests) with auto-shutoff.
  • One-button calibration; typical calibration interval two weeks to monthly.
  • Automatic temperature compensation; adjustable or disableable.
  • User-adjustable conductivity/TDS conversion ratio for nonstandard solutions.
  • Data retained in non-volatile memory; waterproof and buoyant-operational after immersion up to 1 m (3 ft).