Learn More
Hach Company Hach Online Data
Catalog No. 5000207117 Shop All Hach Company Products
Encompass_Preferred
This software package includes an USB cable and a Setup USB-stick The Setup installs a MS Windows device driver (DLL) for Hach Spectrophotometers DR3900 DR6000 and the Lico spectral colorimeter as well as a Microsoft Excel template file with programmed macro functions for reading and displaying the photometric measurement data captured via USB connection
Catalog No. 50-002-07117 Supplier Hach Company Supplier No. LZV799
This software package includes an USB cable and a Setup USB-stick The Setup installs a MS Windows device driver (DLL) for Hach Spectrophotometers DR3900 DR6000 and the Lico spectral colorimeter as well as a Microsoft Excel template file with programmed macro functions for reading and displaying the photometric measurement data captured via USB connection