Learn More
Hach Company Hach Online Process ORP Sensor, General Purpose Analog, PEEK housing, 15 m cable (RD1P5A50)
Catalog No. 5000207591 Shop All Hach Company Products
Encompass
The Hach Online Process ORP Sensor (analog) provides accurate, reliable oxidation-reduction potential measurement for process water and wastewater applications using differential electrode measurement and a replaceable double-junction salt bridge to reduce contamination and extend calibration intervals.
  • General-purpose platinum ORP electrode suitable for a wide range of process conditions.
  • PEEK housing provides chemical resistance and durability.
  • Three-electrode differential measurement reduces reference junction potential and eliminates sensor ground loops.
  • Replaceable double-junction salt bridge protects the reference electrode and extends sensor life.
  • Encapsulated preamplifier supports sensor placement up to 1000 m (3280 ft) from the analyzer.
  • Includes a 15 m (50 ft) cable and analog cable connection for conventional controllers.
Catalog No. 50-002-07591 Supplier Hach Company Supplier No. RD1P5A50
The Hach Online Process ORP Sensor (analog) provides accurate, reliable oxidation-reduction potential measurement for process water and wastewater applications using differential electrode measurement and a replaceable double-junction salt bridge to reduce contamination and extend calibration intervals.
  • General-purpose platinum ORP electrode suitable for a wide range of process conditions.
  • PEEK housing provides chemical resistance and durability.
  • Three-electrode differential measurement reduces reference junction potential and eliminates sensor ground loops.
  • Replaceable double-junction salt bridge protects the reference electrode and extends sensor life.
  • Encapsulated preamplifier supports sensor placement up to 1000 m (3280 ft) from the analyzer.
  • Includes a 15 m (50 ft) cable and analog cable connection for conventional controllers.