Learn More
Hach Company Hach pH sensor, convertible mounting, PPS body, low-pH glass electrode, 5-wire built-in preamplifier, 10 ft cable
Catalog No. 5000207707 Shop All Hach Company Products
Encompass
The Hach pH sensor is a convertible-mounting industrial probe with a PPS body, low-pH glass electrode, and an internal preamplifier. It provides reliable differential pH measurement for process and water applications and includes a 10 ft (3 m) cable for straightforward installation.
  • Convertible mounting for immersion or inline/tee installation.
  • PPS body for chemical resistance and durability.
  • Low-pH glass differential electrode for accurate measurements.
  • PVDF reference junction and replaceable salt bridge to reduce contamination.
  • 5-wire configuration with built-in preamplifier for long-distance signal transmission.
  • Includes 10 ft (3 m) cable; supports analyzer placement up to 3,000 ft (914 m).
  • 12-month warranty.
Catalog No. 50-002-07707 Supplier Hach Company Supplier No. 6022P610
The Hach pH sensor is a convertible-mounting industrial probe with a PPS body, low-pH glass electrode, and an internal preamplifier. It provides reliable differential pH measurement for process and water applications and includes a 10 ft (3 m) cable for straightforward installation.
  • Convertible mounting for immersion or inline/tee installation.
  • PPS body for chemical resistance and durability.
  • Low-pH glass differential electrode for accurate measurements.
  • PVDF reference junction and replaceable salt bridge to reduce contamination.
  • 5-wire configuration with built-in preamplifier for long-distance signal transmission.
  • Includes 10 ft (3 m) cable; supports analyzer placement up to 3,000 ft (914 m).
  • 12-month warranty.