Learn More
Hach Company Hach pHD Online Process pH Sensor - General Purpose Insertion-Mount Glass Electrode, PEEK Housing, 30 ft Cable (PD2P1A30)
Catalog No. 5000207282 Shop All Hach Company Products
Encompass
The pHD online process pH sensor is an insertion-mount, general-purpose pH sensor designed for continuous online monitoring in industrial and wastewater process applications. It uses pHD technology with a glass pH electrode in a PEEK housing and includes a factory-attached cable for straightforward installation and analog output for connection to controllers.
  • pHD technology and glass pH electrode for accurate measurements.
  • Insertion-mount design for continuous online monitoring.
  • PEEK housing for chemical resistance and durability.
  • 30 ft factory-attached cable for flexible installation.
  • Analog output for compatibility with controllers and transmitters.
  • Suitable for process control and wastewater monitoring applications.
Catalog No. 50-002-07282 Supplier Hach Company Supplier No. PD2P1A30
The pHD online process pH sensor is an insertion-mount, general-purpose pH sensor designed for continuous online monitoring in industrial and wastewater process applications. It uses pHD technology with a glass pH electrode in a PEEK housing and includes a factory-attached cable for straightforward installation and analog output for connection to controllers.
  • pHD technology and glass pH electrode for accurate measurements.
  • Insertion-mount design for continuous online monitoring.
  • PEEK housing for chemical resistance and durability.
  • 30 ft factory-attached cable for flexible installation.
  • Analog output for compatibility with controllers and transmitters.
  • Suitable for process control and wastewater monitoring applications.