Learn More
Hach Company Hach Ring for 10 mL Syringe for AT/KF Titrator (LZE106)
Supplier:  Hach Company LZE106
Encompass
Replacement ring that secures a 10 mL syringe used in AT/KF titrators. This spare part is intended for routine maintenance and to ensure proper syringe positioning and consistent performance during titration procedures.
  • Secures 10 mL titration syringes in the syringe holder.
  • Helps maintain syringe alignment for consistent titration results.
  • Low-cost replacement for routine maintenance and repair.
  • Simple to install without tools for fast serviceability.
  • Compact and lightweight, suitable for laboratory environments.
Catalog No. 50-002-07159