Learn More
Hach Company Hach Sample Cell, 10 mm Rectangular Glass Cuvette, Pack of 3 (LZP045)
Catalog No. 5000207201 Shop All Hach Company Products
Encompass
This rectangular glass sample cell has a 10 mm optical path length and is supplied as a pack of three. It is designed for use in spectrophotometers, colorimeters, and other optical measurement instruments that require a standard 10 mm pathlength.
  • 10 mm optical path length for standard spectrophotometric measurements.
  • Rectangular geometry compatible with many benchtop colorimeters and spectrometers.
  • Glass construction provides optical clarity and chemical resistance.
  • Pack of three for replacement, spare, or multiple-sample use.
  • Suitable for routine laboratory and field colorimetric analyses.
Catalog No. 50-002-07201 Supplier Hach Company Supplier No. LZP045
This rectangular glass sample cell has a 10 mm optical path length and is supplied as a pack of three. It is designed for use in spectrophotometers, colorimeters, and other optical measurement instruments that require a standard 10 mm pathlength.
  • 10 mm optical path length for standard spectrophotometric measurements.
  • Rectangular geometry compatible with many benchtop colorimeters and spectrometers.
  • Glass construction provides optical clarity and chemical resistance.
  • Pack of three for replacement, spare, or multiple-sample use.
  • Suitable for routine laboratory and field colorimetric analyses.