Learn More
Hach Company Hach SC4500 Controller (LXV525.99P11551) - Prognosys universal controller; 5x mA outputs, supports 2 digital sensors, 100-240 VAC, C1D2; sold without power plug.
Catalog No. 5000207367 Shop All Hach Company Products
Encompass
SC4500 series digital controller for online sensor monitoring and process control. Designed for continuous water-quality measurement, it provides multiple analog outputs, digital sensor support, and flexible software configuration for integration into plant control systems.
  • Provides five 4-20 mA outputs for simultaneous process signals.
  • Supports two digital sensors for direct sensor integration.
  • Operates from 100-240 VAC for global voltage compatibility.
  • Includes Prognosys-based software for configuration and diagnostics.
  • Rated for Class I, Division 2 hazardous locations.
  • Supplied without power plug to allow regional power connection options.
Catalog No. 50-002-07367 Supplier Hach Company Supplier No. LXV525.99P11551
SC4500 series digital controller for online sensor monitoring and process control. Designed for continuous water-quality measurement, it provides multiple analog outputs, digital sensor support, and flexible software configuration for integration into plant control systems.
  • Provides five 4-20 mA outputs for simultaneous process signals.
  • Supports two digital sensors for direct sensor integration.
  • Operates from 100-240 VAC for global voltage compatibility.
  • Includes Prognosys-based software for configuration and diagnostics.
  • Rated for Class I, Division 2 hazardous locations.
  • Supplied without power plug to allow regional power connection options.